Skip to Content
Merck
All Photos(1)

Documents

ABE1943

Sigma-Aldrich

Anti-HOXB13 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Homeobox protein Hox-B13, HOXB13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HOXB13(10481)

General description

Homeobox protein Hox-B13 (UniProt Q92826) is encoded by the HOXB13 (also known as PSGD) gene (Gene ID 10481) in human. HOXB13 is a homeobox gene family member whose expression is restricted to prostate tissues and is highly correlated with that of androgen receptor (AR). Hox-B13 suppresses androgen-stimulated AR activity via direct AR interaction and thereby inhibits androgen-dependent (AD) prostate cancer (PCa) growth under androgen-rich conditions. On the other hand, HOXB13 is also known to be over-expressed in castration-resistant PCa (CRPC), where it plays a dichotomic role in promoting androgen-independent (AI) PCa growth. The zinc level in human prostate is markedly diminished in prostatic epithelial tumors and continues to decline during progression towards AI growth. HOXB13 over-expression upregulates transcription of its target genes SLC30A10 (ZnT10) and SLC30A4 (ZnT4) for zinc output transporters, which are responsible for the reduction in zinc level in CRPC. The diminished intracellular zinc level in turn stimulates NF-κB signaling and promotes invasion and metastasis of CRPC cells.

Specificity

This polyclonal antibody selectively reacts with human HOXB13, but not the homologous HOXA13 or HOXD13.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human HOXB13.

Application

This Anti-HOXB13 Antibody is validated for use in Western Blotting, Immunocytochemistry and Immunohistochemistry for the detection of HOXB13.
Western Blotting Analysis: A representative lot detected HOXB13 expression in human prostate cancer cell lines MDA PCa 2b, LNCaP, and C4-2, but not in prostate cancer DU145, PC3, CWR22Rv-1, or the non-tumorigenic prostate epithelial line P69 (Kim, Y.R., et. al. (2010). Mol Cancer. 9:124).
Immunocytochemistry Analysis: A representative lot detected HOXB13 nuclear expression in MDA PCa 2b and LNCaP, but not PC3 or DU145, human prostate cancer cell lines (Kim, Y.R., et. al. (2010). Mol Cancer. 9:124).
Immunohistochemistry Analysis: A representative lot detected differential HOXB13 expressions among various androgen-dependent (AD) and androgen-independent (AI) human prostate cancer tissues (Kim, Y.R., et. al. (2010). Mol Cancer. 9:124; Kim, Y.R., et. al. (2014). Oncogene. 33(37):4558-4567).

Quality

Evaluated by Western Blotting in LNCaP cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected HOXB13 in 10 µg of LNCaP cell lysate.

Target description

~35 kDa observed

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-HCI, pH 8.0, and 0.1% soidum azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service