Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

9710-OP

Millipore

Xylene Cyanol FF

OmniPur® Grade

Synonym(s):

OmniPur® Xylene Cyanol FF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

blue-green

shipped in

ambient

InChI

1S/C25H28N2O6S2.Na/c1-4-26-23-9-5-19(13-17(23)2)11-12-27-24-10-6-20(14-18(24)3)15-21-7-8-22(34(28,29)30)16-25(21)35(31,32)33;/h5-10,13-16,26H,4,11-12H2,1-3H3,(H,28,29,30)(H,31,32,33);/q;+1/p-1/b20-15+,27-24?;

InChI key

UQGVMAOBDMCVOQ-PULQMKEHSA-M

General description

Tracking dye for DNA gels. Molecular biology grade.

Specifications:
Moisture (H2O): 10% max
Molar absorptivity Em (614 nm, methanol): 30000 min





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service