Skip to Content
Merck
All Photos(1)

Key Documents

539122

Sigma-Aldrich

Protamine Sulfate, Salmon Milt

Sign Into View Organizational & Contract Pricing


About This Item

form

solid

Quality Level

color

white

solubility

1.6 N NH4OH: 2.5 mg/mL
PBS: 25 mg/mL
H2O: 5 mg/mL
2 M HCl: 50 mg/mL

General description

Native protamine sulfate from salmon milt. Neutralizes the anticoagulant activity of heparin.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service