Skip to Content
Merck
All Photos(1)

Key Documents

5.06305

Sigma-Aldrich

SKP2 E3 Ligase Inhibitor III

Synonym(s):

SKP2 E3 Ligase Inhibitor III, 3-(1,3-Benzothiazol-2-yl)-6-ethyl-7-hydroxy-8-(1-piperidinylmethyl)-4H-chromen-4-one, 3-(Benzo[d]thiazol-2-yl)-6-ethyl-7-hydroxy-8-(piperidin-1-ylmethyl)-4H-chromen-4-one, 3-Benzothiazol-2-yl-6-ethyl-7-hydroxy-8-(piperidylmethyl)chromen-4-one, p21/Cip1/CKI/Waf1 Activator III, CRL1SKP2 Inhibitor III, p27/Kip1 Activator III, S-phase Kinase-associated Protein 2 Inhibitor III, SCFSKP2 Inhibitor III, SZL-P1-41, p21/Cip1/CKI/Waf1 Activator III, CRL1SKP2 Inhibitor III, p27/Kip1 Activator III, S-phase Kinase-associated Protein 2 Inhibitor III, SCFSKP2 Inhibitor III, SZL-P1-41, 3-(1,3-Benzothiazol-2-yl)-6-ethyl-7-hydroxy-8-(1-piperidinylmethyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N2O3S
Molecular Weight:
420.52
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable benzothiazolylchromenone compound that disrupts Skp2-Skp1 interaction via direct Skp2 affinity binding and selectively prevents Skp2-, but not Fbw7- or β-TrCP-, Skp1-Cullin-1-F-box (SFC) complex-mediated substrates ubiquitination both in cell-free systems and in cultures, effectively preventing ubiquitination-dependent activation (Akt) and degradation (p21 & p27) of Skp2 substrates with concomitant down-regulation of cellular Skp2 protein level (10 to 20 µM in HEK293T & PC3 cultures). Exhibits antiproliferation activity toward a panel of 8 cancer (IC50 from 1.22 to 10.5 µM), but not normal PNT1A epithelial or IMR90 fibroblast, lines and effectively reduces ALDH+ cancer stem cell (CSC; Cancer Initiating Cell) population in prostate cancer PC3 cultures (10 to 20 µM) by promoting p53-independent senescence and blocking Akt-dependent aerobic glycolysis. Exhibits good pharmacokinetic property (tmax = 1 h; t1/2 >6 h; Cmax >1 µM/plasma and >4 µM/tumor in 200 mm3 PC3 tumor-bearing mice; 80 mg/kg i.p.) and is efficacious in suppressing A549 and PC3 tumor growth in mice (40 to 80 mg/kg/d i.p.) in vivo.
A cell-permeable benzothiazolylchromenone compound that disrupts Skp2-Skp1 interaction via direct Skp2 affinity binding and selectively prevents Skp2-, but not Fbw7- or β-TrCP-, Skp1-Cullin-1-F-box (SFC) complex-mediated substrates ubiquitination, effectively preventing Skp2 substrates activation (Akt) and degradation (p21 & p27) with concomitant down-regulation of cellular Skp2 protein level (10 to 20 µM in HEK293T & PC3 cultures). Exhibits antiproliferation activity toward a panel of 8 cancer (IC50 from 1.22 to 10.5 µM) and reduces ALDH+ cancer stem cell population in PC3 cultures (10 to 20 µM). Exhibits good pharmacokinetic property and is efficacious in suppressing A549 and PC3 tumor growth in mice (40 to 80 mg/kg/d i.p.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Skp2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Chan, C.H., et al. 2013 Cell154, 556.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip E D Chung et al.
Nature communications, 11(1), 1825-1825 (2020-04-15)
Pineoblastoma is a rare pediatric cancer induced by germline mutations in the tumor suppressors RB1 or DICER1. Presence of leptomeningeal metastases is indicative of poor prognosis. Here we report that inactivation of Rb plus p53 via a WAP-Cre transgene, commonly

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service