Skip to Content
Merck
All Photos(1)

Key Documents

400060

Millipore

Anti-Glucose Transporter-1 Rabbit pAb

Synonym(s):

Anti-Glut-1

Sign Into View Organizational & Contract Pricing


About This Item

antibody form

affinity isolated antibody

Quality Level

clone

polyclonal

form

liquid

species reactivity

mouse, human, rabbit, rat

isotype

IgG

General description

Glucose transporter-1 (Glut-1) is a transmembrane protein that facilitates transport of glucose in the cell in an energy-independent fashion. Glut-1 is expressed in many tissues and is the only isoform of the glucose transporter family expressed in mammary gland.
Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~42-45 kDa Glut-1 protein.
Recognizes the ~42-45 kDa GLUT-1 protein. Also recognizes the HepG2-type transporter.

Immunogen

a synthetic peptide corresponding to amino acids near the C-terminus of mouse GLUT-1, conjugated to KLH

Application

ELISA (0.5-1 μg/ml)
Immunoblotting (1-10 μg/ml)
Immunofluorescence (see comments)
Frozen Sections (2-10 μg/ml)
Immunoprecipitation (see comments)

Physical form

In PBS, 0.1% BSA, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Rat brain

Other Notes

The antigen used is 100% homologous to mouse, rabbit, human, bovine, pig, and chicken Glut-1. Also recognizes the Hep G2-type transporter. This antibody is also reported to work for immunofluorescence and immunoprecipitation. Variables associated with ass

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mueckler
European journal of biochemistry, 219(3), 713-725 (1994-02-01)
Facilitative glucose transport is mediated by members of the Glut protein family that belong to a much larger superfamily of 12 transmembrane segment transporters. Six members of the Glut family have been described thus far. These proteins are expressed in
H C Haspel et al.
The Journal of biological chemistry, 263(1), 398-403 (1988-01-05)
Peptides corresponding to amino acid residues 1-12 of the amino terminal and 480-492 of the carboxyl terminal of the deduced sequence of the glucose transporter were synthesized and used to produce site-specific polyclonal antipeptide sera. In a solid-phase radioimmunoassay, antiserum
D S Harris et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(16), 7556-7560 (1992-08-15)
We have examined the expression and cellular location of facilitated glucose transporter proteins (GLUT1, -3, and -5) in a human colonic epithelial cell line (Caco-2) by using peptide-specific antibodies. A differential cellular distribution of these transporters was observed in differentiated
Karen Gertz et al.
BMC neurology, 16(1), 144-144 (2016-08-20)
VE-cadherin is the chief constituent of endothelial adherens junctions. However, the role of VE-cadherin in the pathogenesis of cerebrovascular diseases including brain ischemia has not yet been investigated. VE-cadherin heterozygous (VEC(+/-)) mice and wildtype controls were subjected to transient brain
M J Birnbaum et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(16), 5784-5788 (1986-08-01)
Antibody raised against the human erythrocyte glucose transporter identified a recombinant lambda gt11 bacteriophage in a cDNA library prepared from immunoselected polysomal RNA from adult rat brain. The cDNA predicts a 492-amino acid protein that demonstrates 97.6% identity to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service