Skip to Content
Merck
All Photos(1)

Documents

382187

Sigma-Aldrich

HDAC Inhibitor XXIII, Tubastatin A

The HDAC Inhibitor XXIII, Tubastatin A controls the biological activity of HDAC. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

HDAC Inhibitor XXIII, Tubastatin A, N-Hydroxy-4-(2-methyl-1,2,3,4-tetrahydro-pyrido[4,3-b]indol-5-ylmethyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N3O2
Molecular Weight:
335.40
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

tan

solubility

DMSO: 10 mg/mL
ethanol: 250 μg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable carbazolohydroxamate that acts as a highly potent HDAC6-selective inhibitor (IC50 = 15 nM) with much reduced or no activity against other known HDACs. Effectively prevents neuronal cell death (by ≥95% at 10 µM) upon oxidative stress induction by HCA and selectively induces cellular α-tubulin, but not histone H4, hyperacetylation (2.5 to 5 µM) in primary rat cortical neuron cultures.
A cell-permeable carbazolohydroxamate whose zinc-chelating hydroxamic acid moiety and catalytic channel rim-targeting aromatic cap structure render it a highly potent HDAC6-selective inhibitor (IC50 = 15 nM against recombinant human HDAC6) with much reduced or no activity against HDAC8, HDAC1 (IC50 = 0.854 and 16.4 µM, respectively), and other known HDACs (HDAC2/3/4/5/7/9/10/11; IC50 >30 µM). Shown to selectively block HDAC6-, but not HDAC1-, dependent cellular deacetylation as evidenced by its selective induction (2.5 to 5 µM) of α-tubulin, but not histone H4, hyperacetylation in cultured primary rat cortical neurons. Although failing to block cellular glutathione depletion by homocysteic acid (HCA), Tubastatin A nevertheless is reported to prevent neuronal cell death upon oxidative stress induction by HCA in a dose-dependent manner (≥95% protection at 10 µM), presumably by preventing peroxiredoxins deacetylation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Butler, K.V., et al. 2010. J. Am. Chem. Soc.132, 10842.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service