Skip to Content
Merck
All Photos(1)

Documents

344277

Sigma-Aldrich

Forskolin, 1,9-Dideoxy-, Coleus forskohlii - CAS 64657-18-7 - Calbiochem

Synonym(s):

Forskolin, 1,9-Dideoxy-, Coleus forskohlii - CAS 64657-18-7 - Calbiochem, 7β-Acetoxy-8,13-epoxy-6β-hydroxy-labd-14-en-11-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
UNSPSC Code:
12352200

Assay

≥95% (NMR)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C22H34O5/c1-8-20(5)12-14(24)16-21(6)11-9-10-19(3,4)17(21)15(25)18(26-13(2)23)22(16,7)27-20/h8,15-18,25H,1,9-12H2,2-7H3/t15-,16+,17-,18-,20-,21+,22-/m0/s1

InChI key

ZKZMDXUDDJYAIB-SUCLLAFCSA-N

General description

This product has been discontinued.



Natural product that does not stimulate adenylate cyclase. For use as a negative control (EC50 >1 mM).

A naturally occurring analog of forskolin that does not stimulate adenylate cyclase (EC50 >1000 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A negative control that does not stimulate adenylate cyclase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 4 months at 4°C.

Other Notes

Roscetti, G., et al. 1994. Eur. J. Pharmacol. 269, 17.
White, G., et al. 1992. Brain Res.586, 157.
Tehrani, M.H. 1989. Synapse4, 126.
Seamon, K.B., et al. 1984. Proc. Natl. Acad. Sci. USA 81, 5081.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service