Skip to Content
Merck
All Photos(1)

Key Documents

175600

Sigma-Aldrich

Angiogenin, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 pg/μg Endotoxins (pg/μg Angiogenin)

shipped in

dry ice

storage temp.

−70°C

General description

Recombinant, human angiogenin expressed in E. coli. A 14 kDa protein with angeogenic and ribonucleolytic activities. Enhances the proteolytic and fibrinolytic activities of endothelial cells. Also, stimulates the endothelial cell invasion of Matrigel basement membrane. Has been shown to stimulate capillary and umbilical vein endothelial cells to produce DAG and secrete prostacyclin by phospholipase activation.
Recombinant, human angiogenin expressed in E. coli. Exhibits angiogenic and ribonucleolytic activities. Enhances the ability of endothelial cells to digest extracellular matrix components and degrade basement membrane. Has been shown to stimulate capillary and umbilical vein endothelial cells to produce DAG and secrete prostacyclin by phospholipase activation. Biological activity: 1.0 µg protein produces an absorbance change at 260 nm of approximately 2.0 - 3.0 as measured by yeast tRNA ribonucleolytic activity.

Biochem/physiol Actions

≥1.0 µg protein produces an absorbance change at 260 nm of ~2.0-3.0 as measured based upon its ribonucleolytic activity toward Yeast tRNA.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS containing 50 µg BSA/µg Angiogenin.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to months at -70°C. Avoid freeze/thaw cycles of solutions.
Reconstitute to a concentration ≥1 µg/ml with sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Leland, P.A., et al. 2002. Biochemistry41, 1343.
Hatzi, E., et al. 2000. Biochem. Biophys. Res. Commun. 267, 719.
Hu, G., et al. 1994. Proc. Natl. Acad. Sci. USA91, 12096.
Lee, F.S., and Vallee, B.L. 1989. Biochem. Biophys. Res. Commun.161, 121.
Kurachi, R., et al. 1985. Biochemistry24, 5494.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jürgen Krauss et al.
Methods in molecular biology (Clifton, N.J.), 525, 471-490 (2009-03-03)
ImmunoRNases represent a highly attractive alternative to conventional immunotoxins for cancer therapy. Quantitative production of immunoRNases in appropriate expression systems, however, remains a major challenge for further clinical development of these novel compounds. Here we describe a method for high-level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service