Skip to Content
Merck
All Photos(1)

Key Documents

KEY007387401

Key Organics / BIONET

3-(pyrimidin-2-yloxy)benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25MG
CA$37.90

CA$37.90


Please contact Customer Service for Availability


Select a Size

Change View
25MG
CA$37.90

About This Item

CAS Number:
Molecular Weight:
200.2
MDL number:

CA$37.90


Please contact Customer Service for Availability

mp

108 - 110

Iupac Name

3-(2-pyrimidinyloxy)benzaldehyde

Inchi Code

1S/C11H8N2O2/c14-8-9-3-1-4-10(7-9)15-11-12-5-2-6-13-11/h1-8H

InChI key

DWRSRAYKUJFONM-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10T-0605

Melting Point

108 - 110

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/10T-0605


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service