JWPH3249DC49
J & W Pharmlab LLC
4-Phenyl-3-piperidin-4-yl-oxazolidin-2-one hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-phenyl-3-(piperidin-4-yl)oxazolidin-2-one hydrochloride
Inchi Code
1S/C14H18N2O2.ClH/c17-14-16(12-6-8-15-9-7-12)13(10-18-14)11-4-2-1-3-5-11;/h1-5,12-13,15H,6-10H2;1H
InChI key
XPTYXJVBOMPCIN-UHFFFAOYSA-N
Purity
97%
Country of Origin
US
Physical Form
White Solid
Product Link
https://www.jwpharmlab.com/home/product/60R0934S
MSDS
https://www.jwpharmlab.com/home/product/60R0934S
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service