Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D9E2

J & W Pharmlab LLC

4-Piperidin-4-yl-benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
240.33
MDL number:

Iupac Name

4-(4-piperidinyl)benzenesulfonamide

Inchi Code

1S/C11H16N2O2S/c12-16(14,15)11-3-1-9(2-4-11)10-5-7-13-8-6-10/h1-4,10,13H,5-8H2,(H2,12,14,15)

InChI key

QAKHULNHTKVTLH-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Light Orange Solid

Product Link

https://www.jwpharmlab.com/home/product/60R0187

MSDS

https://www.jwpharmlab.com/home/product/60R0187


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service