Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11E0DBCF

Enamine

tert-butyl N-(3-sulfanylphenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.31
MDL number:

Iupac Name

tert-butyl 3-sulfanylphenylcarbamate

Inchi Code

1S/C11H15NO2S/c1-11(2,3)14-10(13)12-8-5-4-6-9(15)7-8/h4-7,15H,1-3H3,(H,12,13)

InChI key

FKQZBRIRXCOQJC-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1881763

MSDS

https://www.enaminestore.com/catalog/EN300-1881763


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service