Skip to Content
Merck
All Photos(1)

Key Documents

CIAH988503C3

ChemScene LLC (Preferred Partner)

5-Chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
278.48
MDL number:

Pricing and availability is not currently available. Contact Technical Service

bp

415.4+-40.0 C at 760 mmHg

Iupac Name

5-chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine

Inchi Code

1S/C7H4ClIN2/c8-7-1-4-5(9)2-10-6(4)3-11-7/h1-3,10H

InChI key

INXXPKHKUUMVKX-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1190310-51-0.html

Boiling point

415.4+-40.0 C at 760 mmHg

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W022351.pdf


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service