Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CE79D

Chemcia Scientific, LLC

Methyl-thiazol-2-yl-carbamic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.29
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

tert-butyl methyl(thiazol-2-yl)carbamate

Inchi Code

1S/C9H14N2O2S/c1-9(2,3)13-8(12)11(4)7-10-5-6-14-7/h5-6H,1-4H3

InChI key

YEFAMUXMAJVGDW-UHFFFAOYSA-N

storage temp.

0-5C

Purity

96%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

White Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=2452

MSDS

http://www.chemcia.com/product_fl02.asp?id=2452


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service