Skip to Content
Merck
All Photos(1)

Key Documents

860504P

Avanti

C4 Ceramide (d18:1/4:0)

Avanti Research - A Croda Brand 860504P, powder

Synonym(s):

N-butyroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$451.00

R$451.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$451.00

About This Item

Empirical Formula (Hill Notation):
C22H43NO3
CAS Number:
Molecular Weight:
369.58
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$451.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860504P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860504P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C22H43NO3/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-18-21(25)20(19-24)23-22(26)17-4-2/h16,18,20-21,24-25H,3-15,17,19H2,1-2H3,(H,23,26)/b18-16+/t20-,21+/m0/s1

InChI key

UCBLGIBMIAFISC-CQLAPORSSA-N

General description

C4 Ceramide (d18:1/4:0) or N-butyroyl-D-erythro-sphingosine is a synthetic ceramide containing sphingosine with butyric acid. It is a non-toxic, short-chain sphingolipid.[1]

Application

C4 Ceramide (d18:1/4:0) has been used to study its role in rescuing and stabilizing ΔF508-cystic fibrosis transmembrane conductance regulator (ΔF508-CFTR).[1]

Biochem/physiol Actions

C4 Ceramide (d18:1/4:0) is implicated in a series of mutual autophosphorylation and phosphorylation actions between 3-phosphoinositide-dependent kinase 1 (PDK1) and serum/glucocorticoid-induced protein kinase 1 (SGK1). It stimulates cyclic adenosine monophosphate (cAMP)-activated chloride secretion and represses hypersecretion of interleukin-8 (IL-8). C4 Ceramide might be a potential therapeutic for cystic fibrosis (CF).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860504P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung Caohuy et al.
The Journal of biological chemistry, 289(52), 35953-35968 (2014-11-12)
Cystic fibrosis (CF) is due to a folding defect in the CF transmembrane conductance regulator (CFTR) protein. The most common mutation, ΔF508, prevents CFTR from trafficking to the apical plasma membrane. Here we show that activation of the PDK1/SGK1 signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service