Skip to Content
Merck
All Photos(2)

Key Documents

700081P

Avanti

7β,25-dihydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3β,7β,25-triol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,247.00

R$1,247.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
R$1,247.00

About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
UNSPSC Code:
12352211
NACRES:
NA.25

R$1,247.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700081P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(CC[C@H](O)C2)C2=C[C@H](O)C3C1CCC4(C)C3CCC4C(CCCC(C)(O)C)C

InChI key

BQMSKLCEWBSPPY-LVQATTALSA-N

General description

7β,25-dihydroxycholesterol (7β25OHC) is an isomer of 7α,25-dihydroxycholesterol (7α25OHC).[1] Its synthesis from 7-keto,25-hydroxycholesterol (7k25OHC) is catalyzed by 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1). 11β-Hydroxysteroid dehydrogenase type 2 (11β-HSD2) catalyzes the reverse oxidation from 7β25OHC to 7k25OHC.[1]

Application

7β,25-dihydroxycholesterol has been used as a substrate for 11β-Hydroxysteroid dehydrogenase (11β-HSD)-dependent metabolic studies of oxysterols in human embryonic kidney (HEK-293) cells in enzyme kinetics assay[1] and as a standard in high-performance liquid chromatography(HPLC) for detection of dihydroxycholesterols in human plasma.[2]

Biochem/physiol Actions

7β,25-dihydroxycholesterol is an Epstein-Barr virus-induced gene 2 (EBI2) ligand and is comparatively a weaker chemoattractant than 7α,25-dihydroxycholesterol (7α25OHC).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700081P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina R Beck et al.
The Journal of steroid biochemistry and molecular biology, 190, 19-28 (2019-03-25)
Oxysterols are cholesterol metabolites derived through either autoxidation or enzymatic processes. They consist of a large family of bioactive lipids that have been associated with the progression of multiple pathologies. In order to unravel (patho-)physiological mechanisms involving oxysterols, it is
Ratna Karuna et al.
Steroids, 99(Pt B), 131-138 (2015-02-17)
We report a straightforward sample preparation procedure and a direct liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the analysis of 7alpha,25-dihydroxycholesterol (7α25-OHC) and 7alpha,27-dihydroxycholesterol (7α27-OHC). By applying a slow protein precipitation approach using cold ethanol, we were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service