Skip to Content
Merck
All Photos(1)

Key Documents

ATE443544361

AstaTech, Inc.

5-BROMO-1H-PYRROLE-2-CARBOXAMIDE

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
189.01
MDL number:

Iupac Name

5-bromo-1H-pyrrole-2-carboxamide

Inchi Code

1S/C5H5BrN2O/c6-4-2-1-3(8-4)5(7)9/h1-2,8H,(H2,7,9)

InChI key

IUCIXLJBQZTUQS-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=53474

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=38755


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service