Skip to Content
Merck
All Photos(1)

Key Documents

APO428499170

Apollo Scientific Ltd

2-Methyl-5-(pentafluorosulfur)benzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
311.11
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2-(bromomethyl)-1-methyl-4-(pentafluoro-lambda~6~-sulfanyl)benzene

Inchi Code

1S/C8H8BrF5S/c1-6-2-3-8(4-7(6)5-9)15(10,11,12,13)14/h2-4H,5H2,1H3

InChI key

GQWITWAQAPQUDN-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/2-methyl-5-pentafluorosulfurbenzyl-bromide

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-methyl-5-pentafluorosulfurbenzyl-bromide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service