AMBH97F11377
Ambeed, Inc.
2-(Azepan-1-yl)isonicotinic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(1-azepanyl)isonicotinic acid
Inchi Code
1S/C12H16N2O2/c15-12(16)10-5-6-13-11(9-10)14-7-3-1-2-4-8-14/h5-6,9H,1-4,7-8H2,(H,15,16)
InChI key
UYWSWXXMUJAYFS-UHFFFAOYSA-N
storage temp.
Sealed in dry,Room Temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/885277-05-4.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/885277-05-4.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service