Z269212
Pall membrane filters
Supor membrane, diam. 47 mm, pore size 0.2 μm
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
41104900
eCl@ss:
32110203
Recommended Products
description
Supor membrane
packaging
pkg of 100 ea
manufacturer/tradename
Gelman 60301
diam.
47 mm
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
General description
Offer a wide variety of membranes that meet almost any filtration need.
Supor® polyethersulfone, for biological and pharmaceutical solutions, fast flow, low extractables, autoclavable.
HT Tuffryn® polysulfone, for media and air samples, low-protein-binding, autoclavable.
GN Metricel® mixed cellulose esters, for air samples and microbiological analysis, meets US EPA and ASTM specifications, autoclavable.
Nylaflo™ nylon, for general-purpose applications, HPLC, good chemical resistance, low extractables.
PTFE PTFE on PP support, for aggressive organics, HPLC and air/gases, hydrophobic, chemically inert.
Supor® polyethersulfone, for biological and pharmaceutical solutions, fast flow, low extractables, autoclavable.
HT Tuffryn® polysulfone, for media and air samples, low-protein-binding, autoclavable.
GN Metricel® mixed cellulose esters, for air samples and microbiological analysis, meets US EPA and ASTM specifications, autoclavable.
Nylaflo™ nylon, for general-purpose applications, HPLC, good chemical resistance, low extractables.
PTFE PTFE on PP support, for aggressive organics, HPLC and air/gases, hydrophobic, chemically inert.
Legal Information
Metricel is a registered trademark of Pall Corporation
Nylaflo is a trademark of Pall Corporation
Supor is a registered trademark of Pall Corporation
Tuffryn is a registered trademark of Pall Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service