Key Documents
Z252956
Sigma-Aldrich
Aldrich® Flavors and Fragrances test strips
L × W 5 in. × 3 1/2 in.
Select a Size
100 EA
R$203.00
R$203.00
Estimated to ship onApril 24, 2025
Select a Size
Change View
100 EA
R$203.00
About This Item
UNSPSC Code:
41113034
NACRES:
NB.35
R$203.00
Estimated to ship onApril 24, 2025
packaging
pkg of 100 ea
L × W
5 in. × 3 1/2 in.
General description
Dip and sniff test strips for sampling aroma chemicals.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)