Skip to Content
Merck
All Photos(1)

Documents

W206806

Sigma-Aldrich

Amyl formate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2(CH2)4CH3
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2068
EC Number:
Council of Europe no.:
497
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.159
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.399 (lit.)

bp

132 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; earthy; fruity

SMILES string

[H]C(=O)OCCCCC

InChI

1S/C6H12O2/c1-2-3-4-5-8-6-7/h6H,2-5H2,1H3

InChI key

DIQMPQMYFZXDAX-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service