Skip to Content
Merck
All Photos(1)

Documents

M30604

Sigma-Aldrich

2-Methylbenzoselenazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NSe
CAS Number:
Molecular Weight:
196.11
Beilstein:
112836
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.652 (lit.)

bp

140 °C/51 mmHg (lit.)

mp

28.5-30 °C (lit.)

SMILES string

Cc1nc2ccccc2[se]1

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service