GF04159245
T1 - thermocouple alloy, Ni90Cr10
insulated wire, 100m length, 0.125mm conductor diameter, 0.015mm insulation thickness, polyester insulation
Synonym(s):
NI095840
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
polyester insulation
thermocouple wire
form
wire (insulated)
manufacturer/tradename
Goodfellow 041-592-45
insulation thickness
0.015 mm
wire L × diam.
100 m × 0.125 mm , conductor diameter
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Application
may be used as one component of a thermocouple
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service