DWK211836309
Duran® Filtering Flasks And Bottles With Side-Arm Socket
conical bottom (ERLENMEYER SHAPE)
Synonym(s):
211836309
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
conical bottom (ERLENMEYER SHAPE)
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 211836309
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
Application
Separations by vacuum filtration, Filtration at reduced pressure, Filtrieren, Vacuum filtration, Safe working under vacuum, Boro 3.6
Features and Benefits
Provision of filtration flasks with a socket has not only made work in preparation and analytical laboratories easier and simpler, but has also reduced the risk of accidents; Vacuum resistant due to the heavy wall thickness; Fired-on, highly durable white print; Fired-on, highly durable white print; Erlenmeyer shape
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service