Skip to Content
Merck
All Photos(1)

Documents

933279

Sigma-Aldrich

SEM Sample Holder

Variable Tilt Mount holder, For Pin

Synonym(s):

variable tilt mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware
The mount is marked for 30°, 45°, 70° and 90°. The table measures 11 x 11 mm (0.46" x0.46")and the overall size is 14 x 14 x12.7 mm.

Application

This mount which is made from aluminum is used for the mounting and tilting of samples from 0° – 90° as well as for SEM investigation with small working distances.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service