Skip to Content
Merck
All Photos(3)

Key Documents

900977

Sigma-Aldrich

SPPO13

Synonym(s):

1,1′-(9,9′-Spirobi[9H-fluorene]-2,7-diyl)bis[1,1-diphenylphosphine oxide], 2,7-Bis(diphenylphosphoryl)-9,9′-spirobifluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H34O2P2
CAS Number:
Molecular Weight:
716.74
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

powder or crystals

SMILES string

[P](=O)(c%10ccccc%10)(c9ccccc9)c1cc2c(cc1)c3c(cc(cc3)[P](=O)(c8ccccc8)c7ccccc7)C42c5c(cccc5)c6c4cccc6

InChI

1S/C49H34O2P2/c50-52(35-17-5-1-6-18-35,36-19-7-2-8-20-36)39-29-31-43-44-32-30-40(53(51,37-21-9-3-10-22-37)38-23-11-4-12-24-38)34-48(44)49(47(43)33-39)45-27-15-13-25-41(45)42-26-14-16-28-46(42)49/h1-34H

InChI key

QZSXXAOHMOQXAZ-UHFFFAOYSA-N

General description

OLED phosphorescent host material.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Wang et al.
ACS applied materials & interfaces, 8(43), 29600-29607 (2016-11-03)
A novel self-host blue-emitting iridium dendrimer, namely, B-CzPO, has been designed and synthesized via a postdendronization route, where a bipolar carbazole/triphenylphosphine oxide hybrid is selected as the peripheral dendron instead of the p-type oligocarbazole used in unipolar analogue B-CzG2. This
A novel blue fluorescent polymer for solution-processed fluorescent-phosphorescent hybrid WOLEDs.
Wang J, et al.
Journal of Material Chemistry C, 3(12), 2856-2864 (2015)
Fred Ka-Wai Kong et al.
Journal of the American Chemical Society, 138(19), 6281-6291 (2016-05-05)
A new class of luminescent dendritic carbazole-containing alkynylplatinum(II) complexes has been synthesized, characterized, and applied as phosphorescent dopants in the fabrication of solution-processable organic light-emitting devices (OLEDs). These complexes exhibit high photoluminescence quantum yields of up to 80% in spin-coated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service