Skip to Content
Merck
All Photos(1)

Key Documents

862215

Sigma-Aldrich

S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine

97%

Synonym(s):

6-[(2-Hydroxy-5-nitrobenzyl)thio]-9-β-D-ribofuranosylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N5O7S
CAS Number:
Molecular Weight:
435.41
MDL number:
UNSPSC Code:
41100000
PubChem Substance ID:

Assay

97%

SMILES string

OC[C@@H]1O[C@@H]([C@@H](O)[C@H]1O)n2cnc3c(SCc4cc(ccc4O)[N+]([O-])=O)ncnc23

InChI

1S/C17H17N5O7S/c23-4-11-13(25)14(26)17(29-11)21-7-20-12-15(21)18-6-19-16(12)30-5-8-3-9(22(27)28)1-2-10(8)24/h1-3,6-7,11,13-14,17,23-26H,4-5H2/t11-,13-,14-,17-/m1/s1

InChI key

UNTSTPIAQOSGHD-LSCFUAHRSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service