Skip to Content
Merck
All Photos(1)

Documents

579408

Sigma-Aldrich

1-(Methylsulfonyl)-1H-benzotriazole

95%

Synonym(s):

1-Methanesulfonyl-1H-benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3O2S
CAS Number:
Molecular Weight:
197.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

108-112 °C (lit.)

SMILES string

CS(=O)(=O)n1nnc2ccccc12

InChI

1S/C7H7N3O2S/c1-13(11,12)10-7-5-3-2-4-6(7)8-9-10/h2-5H,1H3

InChI key

OMMZSDGGKVRGGP-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed Elagawany et al.
Biomedical chromatography : BMC, 33(9), e4579-e4579 (2019-05-12)
An efficient, economic and high yielding method was described for the synthesis of baclofen (BAC) pharmacopoeial impurities (impurity A and impurity B) which can be used for gram-scale synthesis. Furthermore, a novel ecofriendly thin-layer chromatographic TLC-densitometric method was established and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service