Recommended Products
Assay
95% (GC)
mp
86-91 °C (lit.)
SMILES string
COC(=O)c1ccc(N)c(I)c1
InChI
1S/C8H8INO2/c1-12-8(11)5-2-3-7(10)6(9)4-5/h2-4H,10H2,1H3
InChI key
MRLVFVTVXSKAMX-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pyrrolidinobenzoic acid inhibitors of influenza virus neuraminidase: The hydrophobic side chain influences type A subtype selectivity.
Bioorganic & Medicinal Chemistry, 20(14), 4582-4589 (2012)
Targeting aldehyde dehydrogenase: a potential approach for cell labeling.
Nuclear Medicine and Biology, 36(8), 919-929 (2009)
Solid-phase synthesis of an AB loop mimetic of the Ce3 domain of human IgE: Macrocyclization by Sonogashira coupling.
The Journal of Organic Chemistry, 68(5), 1843-1851 (2003)
C-N bond formation via Cu-catalyzed cross-coupling with boronic acids leading to methyl carbazole-3-carboxylate: synthesis of carbazole alkaloids.
Royal Society of Chemistry Advances, 4(10), 4960-4969 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service