Skip to Content
Merck
All Photos(1)

Documents

493996

Sigma-Aldrich

2-Chloro-6-methoxy-3-quinolinecarboxaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8ClNO2
CAS Number:
Molecular Weight:
221.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

149-151 °C (lit.)

SMILES string

[H]C(=O)c1cc2cc(OC)ccc2nc1Cl

InChI

1S/C11H8ClNO2/c1-15-9-2-3-10-7(5-9)4-8(6-14)11(12)13-10/h2-6H,1H3

InChI key

TZQOMBXDCIPJKW-UHFFFAOYSA-N

General description

2-Chloro-6-methoxy-3-quinolinecarboxaldehyde is a substituted quinolinecarboxaldehyde.

Application

2-Chloro-6-methoxy-3-quinolinecarboxaldehyde may be used in the preparation of 6-methoxy-2-phenethylquinoline-3-carboxaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajasekhar Reddy Naredla et al.
Tetrahedron, 69(9), 2137-2141 (2013-10-22)
Several phenylethyl-substituted pyridinecarboxaldehydes were prepared from 2-bromo-3-pyridinecarboxaldehyde and these substances are found to undergo cyclization reactions in acidic media. In the absence of added nucleophile, acid promoted cyclization and oxidation (MnO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service