Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457442

Sigma-Aldrich

(E)-5-(2-Bromovinyl)uracil

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrN2O2
CAS Number:
Molecular Weight:
217.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

97%

form

solid

mp

189-194 °C (dec.) (lit.)

functional group

bromo

SMILES string

Br\C=C\C1=CNC(=O)NC1=O

InChI

1S/C6H5BrN2O2/c7-2-1-4-3-8-6(11)9-5(4)10/h1-3H,(H2,8,9,10,11)/b2-1+

InChI key

BLXGZIDBSXVMLU-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Nakayama et al.
Pharmacogenetics, 7(1), 35-43 (1997-02-01)
Sorivudine, 1-beta-D-arabinofuranosyl-5-(E)-(2-bromovinyl)uracil, is a potent antiviral agent against varicella-zoster virus and herpes simplex virus type 1. However, sorivudine should not be used in combination with anticancer drugs such as 5-fluorouracil (5-FU) because (E)-5-(2-bromovinyl)uracil (BVU), a metabolite of sorivudine, inhibits the
Jan Balzarini et al.
Molecular pharmacology, 61(5), 1140-1145 (2002-04-19)
The susceptibility of the bicyclic nucleoside analogs (BCNAs), highly potent and selective inhibitors of varicella-zoster virus (VZV), to the enzymes involved in nucleoside/nucleobase catabolism has been investigated in comparison with the established anti-VZV agent (E)-5-(2-bromovinyl)-2'-deoxyuridine [BVDU; brivudine (Zostex)]. Whereas human
A S Jones et al.
Journal of medicinal chemistry, 24(6), 759-760 (1981-06-01)
(Z)-5-(2-Bromovinyl)uracil was obtained by photoisomerization of the E. isomer. Similarly, (E)-5-(2-bromovinyl)-2'-deoxyuridine gave the required Z isomer. (Z)-5-(2-Bromovinyl)-2'-deoxyuridine is much less active against herpes simplex virus type 1 (HSV-1) and somewhat less active against herpes simplex virus type 2 than is
J Yan et al.
Clinical pharmacology and therapeutics, 61(5), 563-573 (1997-05-01)
Bromovinyl-uracil (BVU) is the principal metabolite of sorivudine, a potent anti-zoster nucleoside. BVU binds to, and irreversibly inhibits, the enzyme dihydropyrimidine dehydrogenase (DPD). The objective of this study was to assess the time course of recovery of DPD activity after
S I Kanamitsu et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(4), 467-474 (2000-03-22)
The fatal drug-drug interaction between sorivudine, an antiviral drug, and 5-fluorouracil (5-FU) has been shown to be caused by a mechanism-based inhibition. In this interaction, sorivudine is converted by gut flora to (E)-5-(2-bromovinyl)uracil (BVU), which is metabolically activated by dihydropyrimidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service