Skip to Content
Merck
All Photos(1)

Key Documents

444693

Sigma-Aldrich

Triethyl 1,3,5-benzenetricarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CO2C2H5)3
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

134-139 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc(cc(c1)C(=O)OCC)C(=O)OCC

InChI

1S/C15H18O6/c1-4-19-13(16)10-7-11(14(17)20-5-2)9-12(8-10)15(18)21-6-3/h7-9H,4-6H2,1-3H3

InChI key

KXGOWZRHSOJOLF-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha Y Chong et al.
Acta crystallographica. Section B, Structural science, 62(Pt 5), 864-874 (2006-09-20)
The crystal structures of triethyl-1,3,5-triazine-2,4,6-tricarboxylate (I), triethyl-1,3,5-benzenetricarboxylate (II) and tris-2-hydroxyethyl isocyanurate (III) have been determined from conventional laboratory X-ray powder diffraction data using the differential evolution structure solution technique. The determination of these structures presented an unexpectedly wide variation in
Synthesis of Macrocyclic [n. n. n](1, 3, 5) Cyclophane Polylactones.
Kanishi M, et al.
Bulletin of the Chemical Society of Japan, 54(12), 3828-3831 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service