Skip to Content
Merck
All Photos(1)

Documents

399272

Sigma-Aldrich

3-Fluorophenethyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2OH
CAS Number:
Molecular Weight:
140.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

90 °C/3 mmHg (lit.)

density

1.125 g/mL at 25 °C (lit.)

SMILES string

OCCc1cccc(F)c1

InChI

1S/C8H9FO/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6,10H,4-5H2

InChI key

MZNBGEKFZCWVES-UHFFFAOYSA-N

General description

3-Fluorophenethyl alcohol is a fluorinated β-phenethyl alcohol. Regioselective synthesis of 3-fluorophenethyl alcohol via regioselective hydroboration has been reported.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective hydroboration-oxidation and-amination of fluoro-substituted styrenes.
Ramachandran PV, et al.
Journal of Fluorine Chemistry, 127(9), 1252-1255 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service