Skip to Content
Merck
All Photos(1)

Key Documents

376922

Sigma-Aldrich

Antimony(III) butoxide solution

65 wt. % in toluene

Synonym(s):

Tributylantimonite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb(OC4H9)3
CAS Number:
Molecular Weight:
341.10
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

1.1 mmHg ( 54.4 °C)

reaction suitability

core: antimony

concentration

65 wt. % in toluene

refractive index

n20/D 1.48

density

1.225 g/mL at 25 °C

SMILES string

CCCCO[Sb](OCCCC)OCCCC

InChI

1S/3C4H9O.Sb/c3*1-2-3-4-5;/h3*2-4H2,1H3;/q3*-1;+3

InChI key

YGBFTDQFAKDXBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used in formation studies of poly(ethylene terephthalate).

Features and Benefits

Reagent used in formation studies of poly(ethylene terephthalate).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Polymer Science Part A: Polymer Chemistry, 26, 2199-2199 (1988)
Journal of Polymer Science Part A: Polymer Chemistry, 27, 3535-3535 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service