Skip to Content
Merck
All Photos(1)

Key Documents

286451

Sigma-Aldrich

Methyl γ-linolenate

98%

Synonym(s):

γ-Linolenic acid methyl ester, ω-6 Linolenic acid methyl ester, 6,9,12-Octadecatrienoic acid methyl ester, Fame 18:3n-6, Methyl (Z,Z,Z)-6,9,12-octadecatrienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)4CO2CH3
CAS Number:
Molecular Weight:
292.46
MDL number:
UNSPSC Code:
12352103

Assay

98%

refractive index

n20/D 1.476 (lit.)

bp

162 °C/0.5 mmHg (lit.)

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCC(=O)OC

InChI

1S/C19H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11,13-14H,3-6,9,12,15-18H2,1-2H3/b8-7-,11-10-,14-13-

InChI key

JFRWATCOFCPIBM-JPFHKJGASA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Jubie et al.
BMC complementary and alternative medicine, 15, 263-263 (2015-08-05)
Isolation of methyl gamma linolenate from Spirulina platensis using flash chromatography and its apoptosis inducing effect against human lung carcinoma A- 549 cell lines. Gamma linolenic acid is an important omega-6 polyunsaturated fatty acid (PUFA) of medicinal interest was isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service