Skip to Content
Merck
All Photos(1)

Key Documents

238678

Sigma-Aldrich

IGEPAL® CO-890

average Mn ~1,982

Synonym(s):

Polyoxyethylene (40) nonylphenyl ether, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n · C15H24O n=40
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~1,982

mp

46-47 °C (lit.)

HLB

17

SMILES string

CCCCCCCCCc1ccc(OCCOCCO)cc1

Legal Information

IGEPAL is a registered trademark of Solvay

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

380.3 °F - closed cup

Flash Point(C)

193.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirko Trilling et al.
Journal of virology, 83(8), 3684-3695 (2009-02-13)
Vaccinia virus (VACV) replicates in mouse and human fibroblasts with comparable kinetics and efficiency, yielding similar titers of infectious progeny. Here we demonstrate that gamma interferon (IFN-gamma) but not IFN-alpha or IFN-beta pretreatment of mouse fibroblasts prior to VACV infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service