Skip to Content
Merck
All Photos(1)

Key Documents

221538

Sigma-Aldrich

1-Pentanesulfonic acid sodium salt

98%

Synonym(s):

Sodium pentanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4SO3Na
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12161900

Assay

98%

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCS([O-])(=O)=O

InChI

1S/C5H12O3S.Na/c1-2-3-4-5-9(6,7)8;/h2-5H2,1H3,(H,6,7,8);/q;+1/p-1

InChI key

ROBLTDOHDSGGDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used as a surfactant in studies on drug release from the water-in-oil-in-water multiple emulsion in vitro.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical & Pharmaceutical Bulletin, 41, 741-741 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service