Skip to Content
Merck
All Photos(1)

Documents

143413

Sigma-Aldrich

1,3-Diaminoguanidine monohydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHC(=NH)NHNH2·HCl
CAS Number:
Molecular Weight:
125.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

180-182 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

Cl.NNC(=N)NN

InChI

1S/CH7N5.ClH/c2-1(5-3)6-4;/h3-4H2,(H3,2,5,6);1H

InChI key

HAZRIBSLCUYMQP-UHFFFAOYSA-N

General description

1,3-Diaminoguanidine monohydrochloride undergoes condensation reaction with:
  • 4-isothiocyanato-4-methylpentane-2-one to yield condensed pyrimidines
  • various aldehydes and ketones to yield bis guanidine derivatives

Other Notes

Contains sodium chloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Brimnes et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 107(6), 555-565 (1999-06-24)
Several N-N-and N-O-containing compounds were analysed for their ability to act as substrates for horseradish peroxidase and peroxidases in Mycobacterium tuberculosis extracts. Aminoguanidine, diaminoguanidine, isoniazid, hydroxylamine and hydrazine were found to be weak substrates for horseradish peroxidase in reaction I
Synthesis of condensed pyrimidines and their evaluation for anti-inflammatory and analgesic activities.
Sondhi SM, et al.
Indian J. Chem. B, 47(1), 136-136 (2008)
Synthesis of biologically active novel bis Schiff bases, bis hydrazone and bis guanidine derivatives.
Sondhi SM, et al.
Indian J. Chem. B, 48(8), 1128-1128 (2009)
Anthony J Lee et al.
Chemical research in toxicology, 18(12), 1927-1933 (2005-12-20)
It is established that aminoguanidine (AG), diaminoguanidine (DAG), and NG-amino-l-arginine (NAA) are metabolism-based inactivators of the three major isoforms of nitric oxide synthase (NOS). In the case of neuronal NOS (nNOS), heme alteration is known to be a major cause
D Dvornik et al.
Journal of diabetes and its complications, 10(1), 23-30 (1996-01-01)
Aminoguanidine, nucleophilic hydrazine derivative, has been shown to inhibit diamine oxidase, the formation of advanced glycation endproducts, nitric oxide synthase, and catalase. Prompted by the reports that aminoguanidine also inhibits aldose reductase (AR), we have investigated the effect of aminoguanidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service