Skip to Content
Merck
All Photos(1)

Documents

118362

Sigma-Aldrich

cis-1,3-Dimethylcyclohexane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

40.5 mmHg ( 37.7 °C)

Assay

99%

autoignition temp.

583 °F

refractive index

n20/D 1.423 (lit.)

bp

120 °C (lit.)

density

0.784 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CCC[C@H](C)C1

InChI

1S/C8H16/c1-7-4-3-5-8(2)6-7/h7-8H,3-6H2,1-2H3/t7-,8+

InChI key

SGVUHPSBDNVHKL-OCAPTIKFSA-N

General description

cis-1,3-Dimethylcyclohexane undergoes ring opening reaction via dicarbene mechanism in the presence of iridium catalyst supported on SiO2.

Application

cis-1,3-Dimethylcyclohexane was used to study the transport and structural phase dynamics exhibited by mixed hydrocarbon multilayer assemblies.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface-mediated segregation and transport processes in mixed hydrocarbon multilayer assemblies.
Bishop AR, et al.
The Journal of Physical Chemistry B, 104(4), 747-753 (2000)
Ring opening of 1, 2-and 1, 3-dimethylcyclohexane on iridium catalysts.
Do PT, et al.
J. Catal., 238(2), 477-488 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service