Key Documents
B2529
Sigma-Aldrich
Semi-dry blotter
Complete unit
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
availability
not available in EU
General description
Complete with two Mylar shields, safety-tip power leads and interlocking safety cover.
Excellent for Western, Northern, or Southern transfers, unit accepts one or several gels up to 20 × 20 cm, and varying thickness.
Stainless steel cathode and platinum anode plates produce highly uniform field and resist corrosion. Mylar shield provided to mask unused area of cathode, prevents arcing, ensures all current passes through gel.
Visit the Document Library
Naresh K Rajendran et al.
Oxidative medicine and cellular longevity, 2021, 6664627-6664627 (2021-01-29)
Increasing evidence suggests that adipose-derived stem cells (ADSCs) serve as a therapeutic approach for wound healing. The aim of this study was to determine the effect of photobiomodulation (PBM) on antioxidant enzymes in ADSCs. Four ADSC cell models, namely, normal