Compelling PlexAbility
Introducing the MILLIPLEX® PLEXpedition 115-Plex Screening Panel
A Fit for Purpose Screening Assay Solution
Simplify your protein biomarker discovery with seamless screening. The MILLIPLEX® PLEXpedition Screening Panel is a versatile and configurable multiplex immunoassay kit designed for the simultaneous screening of up to 115 analytes, including:
- Cytokines, chemokines, and growth factors
- Matrix metalloproteinases
- Biomarkers related to bone health, metabolism, and cardiovascular disease
This panel enables the identification of relative changes in protein expression and is provided in a matrix-free format, allowing for analyte detection in various sample types. Upon identifying markers of interest using this screening panel, you can seamlessly transition to our Qualified MILLIPLEX® panels, which offer optimized serum matrices and lot-to-lot verification for reproducible quantification of protein concentration.
By streamlining the biomarker discovery process, the MILLIPLEX® PLEXpedition Screening Panel accelerates your translational and pre-clinical research while saving you time, money, and sample without sacrificing specificity.
Choose the Analytes You Need Based on the Luminex® Instrument You Have
The MILLIPLEX® PLEXpedition Screening Panel works with all the Luminex® multiplexing instrument systems. However, some instruments can only acquire and analyze data for certain analytes based on their assigned bead region. See the table below for instrument requirements based on analyte selection.
For Research Use Only. Not For Use In Diagnostic Procedures.
Related Resources
- Article: How to Select Your Immunoassay Platform
Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.
- Article: Luminex® Multiplexing Platforms Comparison
Uncover which system is right for your research by comparing the Luminex® multiplexing platforms including the Luminex® 200™, FLEXMAP 3D®, and xMAP® INTELLIFLEX® platforms.
- Article: Overview of Luminex® Multiplex Assay Technology
Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results. See how these multiplex biomarker Luminex®assays are being used to advance research for immunology, cancer, veterinary medicine, metabolism/endocrinology, and more.