Skip to Content
BO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
The benchmark of
The benchmark of
Physical chemistry chemical physics : PCCP (2017-11-25)
Jiří Fukal, Ondřej Páv, Miloš Buděšínský, Jakub Šebera, Vladimír Sychrovský
PMID
29171602
ABSTRACT
A benchmark for structural interpretation of the
Product Description
541508
Sigma-Aldrich
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide, 95%
Pricing
86600
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Pricing