Merck
Human Arylamine
Molecular pharmacology (2017-07-05)
Ximing Xu, Cécile Mathieu, Jérémy Berthelet, Romain Duval, Linh Chi Bui, Florent Busi, Jean-Marie Dupret, Fernando Rodrigues-Lima
ABSTRACT
Thiram (tetramethylthiuram disulfide) is a representative dithiocarbamate (DTC) pesticide used in both the field and as a seed protectant. The widespread use of Thiram and other DTC pesticides has raised concerns for health, because these compounds can exert neuropathic, endocrine disruptive, and carcinogenic effects. These toxic effects are thought to rely, at least in part, on the reaction of Thiram (and certain of its metabolites) with cellular protein thiols with subsequent loss of protein function. So far, a limited number of molecular targets of Thiram have been reported, including few enzymes such as dopamine
Product Description
Supelco
Molinate, PESTANAL®, analytical standard
Supelco
Thiobencarb, PESTANAL®, analytical standard
Supelco
Cycloate, PESTANAL®, analytical standard