Merck
  • Selective targeting of mutant adenomatous polyposis coli (APC) in colorectal cancer.
Selective targeting of mutant adenomatous polyposis coli (APC) in colorectal cancer.
Science translational medicine (2016-11-01)
Lu Zhang, Panayotis C Theodoropoulos, Ugur Eskiocak, Wentian Wang, Young-Ah Moon, Bruce Posner, Noelle S Williams, Woodring E Wright, Sang Bum Kim, Deepak Nijhawan, Jef K De Brabander, Jerry W Shay
ABSTRACT
Mutations in the adenomatous polyposis coli (APC) gene are common in colorectal cancer (CRC), and more than 90% of those mutations generate stable truncated gene products. We describe a chemical screen using normal human colonic epithelial cells (HCECs) and a series of oncogenically progressed HCECs containing a truncated APC protein. With this screen, we identified a small molecule, TASIN-1 (truncated APC selective inhibitor-1), that specifically kills cells with APC truncations but spares normal and cancer cells with wild-type APC. TASIN-1 exerts its cytotoxic effects through inhibition of cholesterol biosynthesis. In vivo administration of TASIN-1 inhibits tumor growth of CRC cells with truncated APC but not APC wild-type CRC cells in xenograft models and in a genetically engineered CRC mouse model with minimal toxicity. TASIN-1 represents a potential therapeutic strategy for prevention and intervention in CRC with mutant APC.
Sigma-Aldrich
TASIN-1 hydrochloride, ≥98% (HPLC)