Merck
  • TDP-43 expression influences amyloidβ plaque deposition and tau aggregation.
TDP-43 expression influences amyloidβ plaque deposition and tau aggregation.
Neurobiology of disease (2017-04-19)
Stephani A Davis, Kok Ann Gan, James A Dowell, Nigel J Cairns, Michael A Gitcho
ABSTRACT
Although the main focus in Alzheimer's disease (AD) has been an investigation of mechanisms causing Aβ plaque deposition and tau tangle formation, recent studies have shown that phosphorylated TDP-43 pathology is present in up to 50% of sporadic cases. Furthermore, elevated phosphorylated TDP-43 has been associated with more severe AD pathology. Therefore, we hypothesized that TDP-43 may regulate amyloid-beta precursor protein (APP) trafficking and tau phosphorylation/aggregation. In order to examine the role of TDP-43 in AD, we developed a transgenic mouse that overexpresses hippocampal and cortical neuronal TDP-43 in a mouse expressing familial mutations (K595N and M596L) in APP and presenilin 1 (PSEN1ΔE9). In our model, increased TDP-43 was related to increased tau aggregation as evidenced by thioflavin S-positive phosphorylated tau, which may implicate TDP-43 expression in pre-tangle formation. In addition, there was increased endosomal/lysosomal localization of APP and reduced Aβ plaque formation with increased TDP-43. Furthermore, there was decreased calcineurin with elevated TDP-43 expression. Since calcineurin is a phosphatase for TDP-43, the decreased calcineurin expression may be one mechanism leading to an increase in accumulation of diffuse phosphorylated TDP-43 in the hippocampus and cortex. We further show that when TDP-43 is knocked down there is an increase in calcineurin. In our model of selective TDP-43 overexpression in an APP/PSEN1 background, we show that TDP-43 decreases Aβ plaque deposition while increasing abnormal tau aggregation. These observations indicate that TDP-43 may play a role in regulating APP trafficking and tau aggregation. Our data suggest that TDP-43 could be a putative target for therapeutic intervention in AD affecting both Aβ plaque formation and tauopathy.
Sigma-Aldrich
Thioflavine S, practical grade
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
MISSION® esiRNA, targeting human TARDBP