Merck
In vitro metabolism of [
Regulatory toxicology and pharmacology : RTP (2016-12-21)
Gopinath C Nallani, Shaaban F ElNaggar, Li Shen, Appavu Chandrasekaran
ABSTRACT
The in vitro comparative animal metabolism study is now a data requirement under EU Directive 1107/2009 for registration of plant protection products. This type of study helps determine the extent of metabolism of a chemical in each surrogate species and whether any unique human metabolite(s) are formed. In the present study, metabolism of racemic [
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Supelco
Benalaxyl, PESTANAL®, analytical standard