Merck
  • Identification of Apo-A1 as a biomarker for early diagnosis of bladder transitional cell carcinoma.
Identification of Apo-A1 as a biomarker for early diagnosis of bladder transitional cell carcinoma.
Proteome science (2011-04-19)
Hongjie Li, Changying Li, Huili Wu, Ting Zhang, Jin Wang, Shixin Wang, Jiwu Chang
ABSTRACT
Bladder transitional cell carcinoma (BTCC) is the fourth most frequent neoplasia in men, clinically characterized by high recurrent rates and poor prognosis. Availability of urinary tumor biomarkers represents a convenient alternative for early detection and disease surveillance because of its direct contact with the tumor and sample accessibility. We tested urine samples from healthy volunteers and patients with low malignant or aggressive BTCC to identify potential biomarkers for early detection of BTCC by two-dimensional electrophoresis (2-DE) coupled with mass spectrometry (MS) and bioinformatics analysis. We observed increased expression of five proteins, including fibrinogen (Fb), lactate dehydrogenase B (LDHB), apolipoprotein-A1 (Apo-A1), clusterin (CLU) and haptoglobin (Hp), which were increased in urine samples of patients with low malignant or aggressive bladder cancer. Further analysis of urine samples of aggressive BTCC showed significant increase in Apo-A1 expression compared to low malignant BTCC. Apo-A1 level was measured quantitatively using enzyme-linked immunosorbent assay (ELISA) and was suggested to provide diagnostic utility to distinguish patients with bladder cancer from controls at 18.22 ng/ml, and distinguish patients with low malignant BTCC from patients with aggressive BTCC in two-tie grading system at 29.86 ng/ml respectively. Further validation assay showed that Apo-A1 could be used as a biomarker to diagnosis BTCC with a sensitivity and specificity of 91.6% and 85.7% respectively, and classify BTCC in two-tie grading system with a sensitivity and specificity of 83.7% and 89.7% respectively. Taken together, our findings suggest Apo-A1 could be a potential biomarker related with early diagnosis and classification in two-tie grading system for bladder cancer.
Sigma-Aldrich
Monoclonal Anti-LDHB antibody produced in mouse, clone 2H6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Fibrinogen antibody produced in mouse, clone FG-21, ascites fluid
Sigma-Aldrich
Monoclonal Anti-CLU antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-APOA1 antibody produced in mouse, clone 3A11-1A9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Haptoglobin antibody produced in mouse, clone HG-36, ascites fluid