Merck
  • Synthesis, structural elucidation and DNA-dependant protein kinase and antiplatelet studies of 2-amino-[5, 6, 7, 8-mono and 7, 8-di-substituted]-1,3-benzoxazines.
Synthesis, structural elucidation and DNA-dependant protein kinase and antiplatelet studies of 2-amino-[5, 6, 7, 8-mono and 7, 8-di-substituted]-1,3-benzoxazines.
European journal of medicinal chemistry (2010-08-31)
Saleh Ihmaid, Jasim Al-Rawi, Christopher Bradley
ABSTRACT
A number of new 2-amino-[5, 6, 7 and 8]-O-substituted 1,3-benzoxazines, and 2-amino 8-methyl-7-O-substituted-1,3-benzoxazines were synthesized. Thirty one new compounds were tested for their effect on collagen induced platelet aggregation and it was found that the most active compounds were 8-methyl-2-morpholin-4-yl-7-(pyridin-3-ylmethoxy)-4H-1,3-benzoxazin-4-one 9f and 8-methyl-2-morpholin-4-yl-7-(pyridin-4-ylmethoxy)-4H-1,3-benzoxazin-4-one 9j with IC(50) = 2 ± 1.5 and 4 ± 2 μM respectively. Inhibition of DNA-PK activity at concentrations of 1.6-4 μM were tested for 9 products 5i, 7a-e and 9b, 9f and 9j.
Sigma-Aldrich
4-(Bromomethyl)pyridine hydrobromide, 97%
Sigma-Aldrich
2-(Bromomethyl)pyridine hydrobromide, 98%