Merck
  • Selective extraction and analysis of catecholamines in rat blood microdialysate by polymeric ionic liquid-diphenylboric acid-packed capillary column and fast separation in high-performance liquid chromatography-electrochemical detector.
Selective extraction and analysis of catecholamines in rat blood microdialysate by polymeric ionic liquid-diphenylboric acid-packed capillary column and fast separation in high-performance liquid chromatography-electrochemical detector.
Journal of chromatography. A (2015-07-25)
Xinguang Zhou, Anwei Zhu, Guoyue Shi
ABSTRACT
Concentration of blood catecholamines (CAs) is linked to a host of cardiovascular diseases, including hypertension and stenocardia. The matrix interferences and low concentration require tedious sample pretreatment methods before quantitative analysis by the gold standard method of high-performance liquid chromatography-electrochemical detector (HPLC-ECD). Solid phase extraction (SPE) has been widely used as the pretreatment technique. Here, a facile polymeric ionic liquid (PIL)-diphenylboric acid (DPBA)-packed capillary column was prepared to selectively extract dopamine (DA), noradrenaline (NE) and epinephrine (E) prior to their quantitative analysis by a fast separation in HPLC-ECD method, while microdialysis sampling method was applied to get the analysis sample. Parameters that influenced desorption efficiency, such as pH, salt concentration, acetonitrile content and wash time, were examined and optimized. The proposed method, combining microdialysis sampling technique, SPE and HPLC-ECD system, was successfully applied to detect CAs in rat blood microdialysate with high sensitivity and selectivity in small sample volumes (5-40μl) and a short analysis time (8min), yielding good reproducibility (RSD 6.5-7.7%) and spiked recovery (91-104.4%).
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), SAJ first grade, ≥98.0%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
DL-4-Hydroxy-3-methoxymandelic acid, ≥98% (HPLC), powder
Sigma-Aldrich
1-Chloro-3-phenylpropane, 99%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%